Avantor® Hichrom HI-5 Basic, GC Columns
Nome del fornitore: Avantor
Informazioni su questo articolo
Max temperature limits may change depending on the film thickness.
24 varianti
Product Details & Documents
Max temperature limits may change depending on the film thickness.
Specifications for Product Family
Specifiche
- Catalog No:
- HI03-25-025-15
- HI03-25-025-30
- HI03-25-025-50
- HI03-25-025-60
- HI03-25-050-15
- HI03-25-050-30
- HI03-25-050-50
- HI03-25-050-60
- HI03-25-100-15
- HI03-25-100-30
- HI03-25-100-50
- HI03-25-100-60
- HI03-32-100-15
- HI03-32-100-30
- HI03-32-100-50
- HI03-32-100-60
- HI03-32-150-15
- HI03-32-150-30
- HI03-32-150-50
- HI03-32-150-60
- HI03-53-100-15
- HI03-53-100-30
- HI03-53-100-50
- HI03-53-100-60
- Phase name:
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- HI-5 Basic
- Phase composition:
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- 5% Phenyl, 95% Methyl Polysiloxane - for basic compounds
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
Specifiche
Product Family Options
Informazioni sul prodotto
- Formato della colonnaSpessore dello stratoØ int.LunghezzaDisponibilitàPrice
- HI03-25-025-15GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-025-15Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-025-30GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-025-30Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-025-50GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-025-50Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-025-60GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-025-60Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-050-15GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-050-15Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-050-30GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-050-30Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-050-50GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-050-50Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-050-60GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-050-60Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-100-15GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-100-15Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-100-30GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-100-30Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-100-50GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-100-50Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-25-100-60GC columnSpecifiche:
More
Specifications
Cat. No.HI03-25-100-60Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-100-15GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-100-15Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-100-30GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-100-30Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-100-50GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-100-50Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-100-60GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-100-60Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-150-15GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-150-15Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-150-30GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-150-30Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-150-50GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-150-50Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-32-150-60GC columnSpecifiche:
More
Specifications
Cat. No.HI03-32-150-60Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-53-100-15GC columnSpecifiche:
More
Specifications
Cat. No.HI03-53-100-15Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-53-100-30GC columnSpecifiche:
More
Specifications
Cat. No.HI03-53-100-30Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-53-100-50GC columnSpecifiche:
More
Specifications
Cat. No.HI03-53-100-50Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes - HI03-53-100-60GC columnSpecifiche:
More
Specifications
Cat. No.HI03-53-100-60Phase nameHI-5 BasicPhase composition5% Phenyl, 95% Methyl Polysiloxane - for basic compoundsPolarityLow-polarity stationary phaseCrossbondYes
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-100-PF-CLog in per disponibilità1 * 100 ST - SFr. 806.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-10-PF-CLog in per disponibilità1 * 10 ST - SFr. 111.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-10-PFY-CLog in per disponibilità1 * 10 ST - SFr. 961.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-1-PFY-CLog in per disponibilità1 * 1 ST - SFr. 99.40
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-25-PF-CLog in per disponibilità1 * 25 ST - SFr. 215.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-2-PFY-CLog in per disponibilità1 * 2 ST - SFr. 199.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-50-PF-CLog in per disponibilità1 * 50 ST - SFr. 414.00
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmLinear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-5-PF-CLog in per disponibilità1 * 5 ST - SFr. 74.10
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mmHI0-5-PFY-CLog in per disponibilità1 * 5 ST - SFr. 491.00