Avantor® Hichrom HI-5 HT, GC Columns
Supplier: Avantor
Certificates
About this item
General purpose column with added temperature stability. For the analysis of solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, pesticides, phenols, etc.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HIGCHI863201015G5
- HI87-25-025-15
- HI87-25-025-30
- HI87-25-015-15
- HI87-25-015-30
- HIGCHI87-25-015-10
- HI87-32-010-30
- HI87-53-025-15
- HI87-53-025-30
- HI87-32-010-10
- HI87-32-010-15
- HI87-53-017-30
- HI87-25-010-15
- HI87-32-010-10G2
- HI87-25-010-30
- HI87-32-015-15
- HI87-10-010-10
- HI87-32-025-30
- HI87-53-010-10
- HI87-32-015-30
- HI87-10-010-5
- HI87-32-025-15
- HI87-53-015-15
- HI87-53-015-30
- HI87-53-010-15
- HI87-53-010-30
- Phase name:
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- HI-5 HT
- Phase composition:
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- 5% Phenyl, 95% Methyl Polysiloxane - high temperature
- Polarity:
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Low-polarity stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Limite di temperatura:
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- 400 °C
- Codice USP:
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41
- G27, G36, G41