Avantor® Hichrom HI-ALC 1, GC Columns
Supplier: Avantor
Certificates
About this item
Application specific column for blood alcohols testing.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI36-32-180-30
- HI36-32-150-30
- HI36-53-300-30
- Phase name:
- HI-ALC 1
- HI-ALC 1
- HI-ALC 1
- Phase composition:
- Proprietary Phase developed for blood alcohol analysis
- Proprietary Phase developed for blood alcohol analysis
- Proprietary Phase developed for blood alcohol analysis
- Crossbond:
- Yes
- Yes
- Yes
- Limite di temperatura:
- 280 °C
- 280 °C
- 280 °C