Orders LinkContactUsLinkComponent
Mop systems, HandySorb
Mop systems, HandySorb
Nome del fornitore:   Brady
Mop systems, HandySorb
Nome del fornitore:  Brady
Certificates
Mop systems, HandySorb
Mop systems, HandySorb
2 varianti
Product Details & Documents
HandySorb is a unique and practical tool to safely remove spills in a highly efficient and easy way.
Safe spill clean-up: works with inert pads or pillows that can be replaced without ever touching the absorbed fluids
Easy to use: offers 1,83 m reach when fully extended and features a no touch snap-on system for pads
Highly efficient: each pad absorbs in seconds up to 25 times its weight
Product Family Options
Informazioni sul prodotto
  • Capacità
    Confezione
    Disponibilità
    Price
  • BRDY150597
    Mop system Basic universal pad refills with mop adapter
    Mop system Basic universal pad refills with mop adapter
    74 L
    100 pads/kit
    Log in per disponibilità
    Log in per disponibilità
    1 * 150 ST
    1 * 150 ST - 469.00 SFr.
    469.00 SFr.
    1 * 150 ST
       469.00 SFr.
  • BRDY150598
    Mop system 'No-Touch' universal pads
    Mop system 'No-Touch' universal pads
    47 L
    25 pads/case
    Log in per disponibilità
    Log in per disponibilità
    1 * 25 ST
    1 * 25 ST - 235.00 SFr.
    235.00 SFr.
    1 * 25 ST
       235.00 SFr.
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..