Absorbent pillows, ALLWICK Universal
104 L, 16 pieces/box, 430×480 mm
Codice catalogo BRDY813792
Nome del fornitore: Brady
Vai a
Product Details & Documents
These absorbent pillows absorb spills and leaking fluids in tight spaces and under persistent drip sources.
- Absorbs a variety of liquids including water, coolants, solvents, oils, antifreeze, etc
- Ideal under machines or in tight spaces
Specifiche
- Capacità:104 L
- L×L:430×480 mm
- Confezione:16 pieces/box