Chemical resistant boot covers, AlphaTec® 2000, Model 406
Catalog # ANSEWH20B-00406-00
Supplier: Ansell
Specifications
- Taglia scarpe:42 - 46
- Applicazioni per abiti:Chemical protection
- Colore:bianco
- Conf:200 PAIR
Specifications
About this item
Superior breathable microporous laminate technology.
- Proven protective barrier to low concentration liquid chemicals, liquid, and particulate biological hazards
- Comfortable moisture vapor permeable fabric helps reduce the risk of heat stress
- Optimised fit, improving wearer comfort and safety
- Low Linting
Boot covers feature tie fastening, elastic to top of boot, and bound seams. Covers are manufactured from non-woven microporous polyethylene laminate material.
Recommended for cleanroom preparation and cleaning, production line support and maintenance, plant and machine maintenance, and veterinary services.
Certifications: Category I