Cleanroom gloves, Microflex® 93-360
Supplier: Ansell
Certificates
About this item
Microflex® 93-360 is a thin, chemical resistant disposable glove designed for cleanroom environments. It is made of an innovate, synthetic composite material (nitrile/neoprene) for tough chemical protection.
- Three layer design for superior protection against harsh chemicals including acids, solvents and bases
- Thin mil construction provides enhanced tactility and dexterity
- Nitrile and neoprene composite prevents risk of Type I allergies
- Easy donning interior provides a dry feel and eases the donning and doffing process
- Non-particulating packaging designed to reduce contamination
- Silicone-free, anti-static
Recommended for: Cleanroom cleaning and preparing; blending, compounding solids and liquids; spill or leakage cleanup; transferring liquids and solids; loading centrifuges and chromatography columns; assembly of parts; weighing and dispensing of solid and liquid raw materials.
Certifications: EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS; EN 421, EN 388 (2000X); Cat. III; AQL 0,65
Packaging: 50 gloves packed in a vacuum-sealed polybag. 10 master polybags inside a polycase liner placed inside a shipper/carton. In total 500 gloves per outer carton.
Specifications
- Catalog No:
- ANSE93-360/5.5-6
- ANSE93-360/10.5-11
- ANSE93-360/9.5-10
- ANSE93-360/6.5-7
- ANSE93-360/8.5-9
- ANSE93-360/7.5-8
- Ambidestro/mano specifico:
- ambidestro
- ambidestro
- ambidestro
- ambidestro
- ambidestro
- ambidestro
- Monouso/riutilizzabile:
- monouso
- monouso
- monouso
- monouso
- monouso
- monouso
- Lattice/senza lattice:
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Senza lattice
- Finitura:
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Textured fingers
- Tipo polsino:
- Cuff_Style-21391099|Cuff_Style-21391103
- Cuff_Style-21391099|Cuff_Style-21391103
- Cuff_Style-21391099|Cuff_Style-21391103
- Cuff_Style-21391099|Cuff_Style-21391103
- Cuff_Style-21391099|Cuff_Style-21391103
- Cuff_Style-21391103|Cuff_Style-21391099
- Spessore delle dita:
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- 0,20 mm
- Spessore palmo:
- 0,198 mm
- 0,198 mm
- 0,198 mm
- 0,198 mm
- 0,198 mm
- 0,198 mm
- Norma EN:
- EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS; EN 421, EN 388 (2000X)
- EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS; EN 421, EN 388 (2000X)
- EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS; EN 421, EN 388 (2000X)
- EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS; EN 421, EN 388 (2000X)
- EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS; EN 421, EN 388 (2000X)
- EN ISO 374-1:2016 Type A (JKLOPST), EN ISO 374-5:2016 VIRUS; EN 421, EN 388 (2000X)
- Antistatico:
- +
- +
- +
- +
- +
- +