Orders LinkContactUsLinkComponent
Antipyrine, Reference Standards
Antipyrine, Reference Standards
Supplier:   USP
Antipyrine, Reference Standards
Supplier:  USP
Certificates
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 1040005.
  • 1040016.
  • Descrizione:
  • Antipyrine
  • Antipyrine Related Compound A
  • Synonym(s):
  • 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one
  • CAS No.:
  • 60-80-0
  • 89-25-8
  • Formula:
  • C₁₁H₁₂N₂O
  • C₁₀H₁₀N₂O
Specifications
Product Family Options
Product Information
  • Descrizione
    Standard
    Tipo di conf.
    Conf
    Availability
    Price
  • Antipyrine
    USP reference standard
    Bottiglia di vetro
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 403.00
    SFr. 403.00
    1 * 200 mg
       SFr. 403.00
    Specifications:
    More
    Specifications
    Cat. No.
    1040005.
    Descrizione
    Antipyrine
    Synonym(s)
    CAS No.
    60-80-0
    Formula
    C₁₁H₁₂N₂O
  • Antipyrine Related Compound A
    USP reference standard
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - SFr. 1’410.00
    SFr. 1’410.00
    1 * 25 mg
       SFr. 1’410.00
    Specifications:
    More
    Specifications
    Cat. No.
    1040016.
    Descrizione
    Antipyrine Related Compound A
    Synonym(s)
    3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one
    CAS No.
    89-25-8
    Formula
    C₁₀H₁₀N₂O