1-(2-Methoxyphenyl)piperazine
Catalog # PROOBP232
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:25 mg
- Seq:2315
Specifications
About this item
:
:192.26 g/mol
Boiling Pt:380 °C (1013 hPa)
Melting Pt:35…38 °C
Density:1.09 g/cm³ (20 °C)
EINECS:252-537-7
UN:2923
ADR:8,III
CAS Number:35386-24-4
Formula:C₁₁H₁₆N₂O
MDL Number:MFCD00005958