DL-Serina
Bottiglia di vetro, 100 g
Catalog # MOLEM61964739
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid
- CAS Number:302-84-1
- Formula:C₃H₇NO₃
- MDL Number:MFCD00064223
- Molecular Weight:105.09 g/mol
- Merck Index:13,08534
- Boiling Pt:395 °C (1013 hPa)
- Melting Pt:243…244 °C
- Density:1,543 g/cm³ (20 °C)
- Storage Temperature:Ambiente
- EINECS:206-130-6
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



