Microscope diamond slide writers
Diamond styluses microscope slide labellers
Catalog # SIMPM795-1
Supplier: Simport Scientific
Jump to:
Product Details & Documents
The microscope diamond slide writer is compatible with a wide variety of slide labellers.
- Consistent even coating of diamond dust
- Stainless steel shaft
This diamond stylus is perfectly suited for the Thermo Scientific PrintMate and MicroWriter, RA Lamb, and TBS Shurmark microscope slide labellers. Manufactured with a consistent even coating of diamond dust.
The 2,35 mm shaft is made from stainless steel.
Specifications
- Descrizione:Diamond styluses microscope slide labellers