Pre-packed columns, 1 and 5 ml, SkillPak™
Informazioni su questo articolo
SkillPak 1 and 5 ml columns are designed for fast method development, resin screening or sample concentration. Pre-packed with TOYOPEARL®, TSKgel® or Ca++Pure-HA™ process chromatography media, these columns offer superior separation of biomelocules, such as monoclonal antibodies, proteins and oligonucleotides. SkillPak 50 and 200 ml pre-packed columns are the best solution for seamless scale up of methods developed on SkillPak 1 and 5.
- Standard fittings (10-32 for 1/16" capillary)
- Max. pressure: 3 bar (1 ml columns and 5 ml TOYOPEARL), ≤4 bar (5 ml TSKgel and Ca++Pure-HA)
SkillPak pre-packed columns are designed for your purification and separation tasks from platform design to pilot scale. These columns are ready to use upon receipt and show excellent physical strength and ideal flow characteristics for industrial downstream process.
These columns guarantee optimal performance and can be operated with commonly used low or media pressure liquid chromatography systems. They are reproducibly packed and take into account the varying compressibility of each resin. This provides an accurate representation of conditions found in full scale columns.
1 ml columns: Shipping buffer of 20% ethanol for TOYOPEARL and TSKgel (with the exception of 0,5 mol/L sodium citrate with 20% ethanol for TSKgel SP-5PW (20) and SuperQ-5PW (20)), 20 mmol/L phosphate with 20% ethanol for Ca++Pure-HA.
5 ml columns: Shipping buffer of 20% ethanol for TOYOPEARL and TSKgel, 20 mmol/L phosphate with 20% ethanol for Ca++Pure-HA. Asymmetry factor (As): 0,8 to 1,4 for TOYOPEARL and TSKgel, 0,8 to 2,6 for Ca++Pure-HA.
63 varianti
Product Details & Documents
SkillPak 1 and 5 ml columns are designed for fast method development, resin screening or sample concentration. Pre-packed with TOYOPEARL®, TSKgel® or Ca++Pure-HA™ process chromatography media, these columns offer superior separation of biomelocules, such as monoclonal antibodies, proteins and oligonucleotides. SkillPak 50 and 200 ml pre-packed columns are the best solution for seamless scale up of methods developed on SkillPak 1 and 5.
- Standard fittings (10-32 for 1/16" capillary)
- Max. pressure: 3 bar (1 ml columns and 5 ml TOYOPEARL), ≤4 bar (5 ml TSKgel and Ca++Pure-HA)
SkillPak pre-packed columns are designed for your purification and separation tasks from platform design to pilot scale. These columns are ready to use upon receipt and show excellent physical strength and ideal flow characteristics for industrial downstream process.
These columns guarantee optimal performance and can be operated with commonly used low or media pressure liquid chromatography systems. They are reproducibly packed and take into account the varying compressibility of each resin. This provides an accurate representation of conditions found in full scale columns.
1 ml columns: Shipping buffer of 20% ethanol for TOYOPEARL and TSKgel (with the exception of 0,5 mol/L sodium citrate with 20% ethanol for TSKgel SP-5PW (20) and SuperQ-5PW (20)), 20 mmol/L phosphate with 20% ethanol for Ca++Pure-HA.
5 ml columns: Shipping buffer of 20% ethanol for TOYOPEARL and TSKgel, 20 mmol/L phosphate with 20% ethanol for Ca++Pure-HA. Asymmetry factor (As): 0,8 to 1,4 for TOYOPEARL and TSKgel, 0,8 to 2,6 for Ca++Pure-HA.
Documents
Certificates
Cerca per numero di lottoTOSOH SkillPak™ Pre-Packed columns, 1 ml and 5 ml
Product Literature
TOYO0045244_TDS_EN
Datasheet
Product Family Options
1 ml columns
Informazioni sul prodotto
- Formato della colonnaDimensioni poriDimensioni particelleØ int.LunghezzaCapacitàVelocità del flussoDisponibilitàPrice
- TOYO0045200TOYOPEARL AF-rProtein L-650FTOYOPEARL AF-rProtein L-650FScouting column100 nm45 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 175.00
- TOYO0045201TOYOPEARL AF-rProtein A HC-650FTOYOPEARL AF-rProtein A HC-650FScouting column100 nm45 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 175.00
- TOYO0045202TOYOPEARL AF-Chelate-650MTOYOPEARL AF-Chelate-650MScouting column100 nm65 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 107.00
- TOYO0045203TOYOPEARL GigaCap Q-650MTOYOPEARL GigaCap Q-650MScouting column100 nm75 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045204TOYOPEARL GigaCap S-650STOYOPEARL GigaCap S-650SScouting column100 nm35 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045205TOYOPEARL Sulfate-650FTOYOPEARL Sulfate-650FScouting column100 nm45 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045206TOYOPEARL SuperQ-650STOYOPEARL SuperQ-650SScouting column100 nm35 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045207TSKgel SP-5PW (20)TSKgel SP-5PW (20)Scouting column100 nm20 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 363.00
- TOYO0045208TSKgel SuperQ-5PW (20)TSKgel SuperQ-5PW (20)Scouting column100 nm20 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 363.00
- TOYO0045209TOYOPEARL NH2-750FTOYOPEARL NH2-750FScouting column> 100 nm45 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045210TOYOPEARL GigaCap DEAE-650MTOYOPEARL GigaCap DEAE-650MScouting column100 nm75 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045211TOYOPEARL GigaCap CM-650MTOYOPEARL GigaCap CM-650MScouting column100 nm75 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045212TOYOPEARL QAE-550CTOYOPEARL QAE-550CScouting column50 nm100 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045213TOYOPEARL SP-550CTOYOPEARL SP-550CScouting column50 nm100 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045214TOYOPEARL Butyl-600MTOYOPEARL Butyl-600MScouting column75 nm65 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045215TOYOPEARL Butyl-650MTOYOPEARL Butyl-650MScouting column100 nm65 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045216TOYOPEARL Phenyl-600MTOYOPEARL Phenyl-600MScouting column75 nm65 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045217TOYOPEARL Phenyl-650MTOYOPEARL Phenyl-650MScouting column100 nm65 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045218TOYOPEARL PPG-600MTOYOPEARL PPG-600MScouting column75 nm65 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045219TOYOPEARL Hexyl-650CTOYOPEARL Hexyl-650CScouting column100 nm100 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045220TOYOPEARL Ether-650MTOYOPEARL Ether-650MScouting column100 nm65 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045221TOYOPEARL AF-rProtein L-650FTOYOPEARL AF-rProtein L-650FScouting column100 nm45 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 862.00
- TOYO0045222TOYOPEARL AF-rProtein A-650FTOYOPEARL AF-rProtein A-650FScouting column100 nm45 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 862.00
- TOYO0045223TOYOPEARL HW-40FTOYOPEARL HW-40FScouting column5 nm45 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045224TOYOPEARL MX-Trp-650MTOYOPEARL MX-Trp-650MScouting column100 nm75 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO0045225Ca++Pure-HACa++Pure-HAScouting column60 - 80 nm39 µm7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 342.00
- TOYO00452262 each (TOYOPEARL GigaCap Q-650M, GigaCap DEAE-650M, NH2-750F)2 each (TOYOPEARL GigaCap Q-650M, GigaCap DEAE-650M, NH2-750F)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 443.00
- TOYO00452272 each (TOYOPEARL Giga-Cap S-650S, GigaCap CM-650M, Sulfate-650F)2 each (TOYOPEARL Giga-Cap S-650S, GigaCap CM-650M, Sulfate-650F)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 443.00
- TOYO00452281 each (TOYOPEARL AF-rProtein A HC-650F, AF-r Protein L-650F, NH2-750F, Sulfate-650F, GigaCapQ-650M GigaCapS-650S)1 each (TOYOPEARL AF-rProtein A HC-650F, AF-r Protein L-650F, NH2-750F, Sulfate-650F, GigaCapQ-650M GigaCapS-650S)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 568.00
- TOYO00452292 each (TOYOPEARL AD-rProtein A HC-650F, Sulfate-650F, NH2-750F)2 each (TOYOPEARL AD-rProtein A HC-650F, Sulfate-650F, NH2-750F)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 568.00
- TOYO00452303 each (TOYOPEARL Sulfate-650F, NH2-750F)3 each (TOYOPEARL Sulfate-650F, NH2-750F)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 443.00
- TOYO00452313 each (Ca++Pure-HA, TOYOPEARL MX-Trp-650M)3 each (Ca++Pure-HA, TOYOPEARL MX-Trp-650M)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 443.00
- TOYO00452321 each (Ca++Pure-HA, TOYOPEARL AF-rProtein A HC-650F, AF-rProtein L-650F, NH2-750F, Sulfate-650F)1 each (Ca++Pure-HA, TOYOPEARL AF-rProtein A HC-650F, AF-rProtein L-650F, NH2-750F, Sulfate-650F)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 561.00
- TOYO00452331 each (TOYOPEARL Butyl-650M, Phenyl-650M, PPG-600M, Hexyl-650C, Ether-650M)1 each (TOYOPEARL Butyl-650M, Phenyl-650M, PPG-600M, Hexyl-650C, Ether-650M)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 369.00
- TOYO00452362 each (TOYOPEARL Butyl-600M, Phenyl-600M, PPG-600M)2 each (TOYOPEARL Butyl-600M, Phenyl-600M, PPG-600M)Kit per lo sviluppo del metodo7,00 mm25 mm1 mlMax. 4 ml/minLog in per disponibilità1 * 1 STSFr. 443.00
5 ml columns
Informazioni sul prodotto
- Formato della colonnaDimensioni poriDimensioni particelleØ int.LunghezzaCapacitàVelocità del flussoDisponibilitàPrice
- TOYO0045239TOYOPEARL GigaCap Q-650MTOYOPEARL GigaCap Q-650MScouting column100 nm75 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045240TOYOPEARL GigaCap S-650STOYOPEARL GigaCap S-650SScouting column100 nm35 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045241TOYOPEARL Sulfate-650FTOYOPEARL Sulfate-650FScouting column100 nm45 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045242TOYOPEARL SuperQ-650STOYOPEARL SuperQ-650SScouting column100 nm35 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045243TSKgel SP-5PW (20)TSKgel SP-5PW (20)Scouting column100 nm20 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 316.00
- TOYO0045244TSKgel SuperQ-5PW (20)TSKgel SuperQ-5PW (20)Scouting column100 nm20 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 316.00
- TOYO0045245TOYOPEARL NH2-750FTOYOPEARL NH2-750FScouting column> 100 nm45 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045246TOYOPEARL GigaCap DEAE-650MTOYOPEARL GigaCap DEAE-650MScouting column100 nm75 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045247TOYOPEARL GigaCap CM-650MTOYOPEARL GigaCap CM-650MScouting column100 nm75 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045248TOYOPEARL QAE-550CTOYOPEARL QAE-550CScouting column50 nm100 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045249TOYOPEARL SP-550CTOYOPEARL SP-550CScouting column50 nm100 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045250TOYOPEARL Butyl-600MTOYOPEARL Butyl-600MScouting column75 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045251TOYOPEARL Butyl-650MTOYOPEARL Butyl-650MScouting column100 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045252TOYOPEARL Phenyl-600MTOYOPEARL Phenyl-600MScouting column75 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045253TOYOPEARL Phenyl-650MTOYOPEARL Phenyl-650MScouting column100 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045254TOYOPEARL PPG-600MTOYOPEARL PPG-600MScouting column75 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045255TOYOPEARL Hexyl-650CTOYOPEARL Hexyl-650CScouting column100 nm100 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045256TOYOPEARL Ether-650MTOYOPEARL Ether-650MScouting column100 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045257TOYOPEARL AF-rProtein L-650FTOYOPEARL AF-rProtein L-650FScouting column100 nm45 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 407.00
- TOYO0045258TOYOPEARL AF-rProtein A HC-650FTOYOPEARL AF-rProtein A HC-650FScouting column100 nm45 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 506.00
- TOYO0045259TOYOPEARL AF-Chelate-650MTOYOPEARL AF-Chelate-650MScouting column100 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 376.00
- TOYO0045260TOYOPEARL HW-40FTOYOPEARL HW-40FScouting column5 nm45 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045261TOYOPEARL MX-Trp-650MTOYOPEARL MX-Trp-650MScouting column100 nm65 µm8,00 mm100 mm5 ml1,3 ml/min (max. 5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO0045262Ca++Pure-HACa++Pure-HAScouting column60 - 80 nm39 µm8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 294.00
- TOYO00452631 each (TOYOPEARL AF-rProtein A HC-650F, Sulfate-650F, NH2-750F)1 each (TOYOPEARL AF-rProtein A HC-650F, Sulfate-650F, NH2-750F)Kit per lo sviluppo del metodo8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 1’120.00
- TOYO00452641 each (TOYOPEARL Sulfat-650F, NH2-750F)1 each (TOYOPEARL Sulfat-650F, NH2-750F)Kit per lo sviluppo del metodo8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 614.00
- TOYO00452651 each (Ca++Pure-HA, TOYOPEARL MX-Trp-650M)1 each (Ca++Pure-HA, TOYOPEARL MX-Trp-650M)Kit per lo sviluppo del metodo8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min Ca++Pure-HA, 5 ml/min TOYOPEARL M)Log in per disponibilità1 * 1 STSFr. 614.00
- TOYO00452661 each (Ca++Pure-HA TOYOPEARL AF-rProtein A HC-650F, AF-rProtein L650F, NH2-750F, Sulfate-650F)1 each (Ca++Pure-HA TOYOPEARL AF-rProtein A HC-650F, AF-rProtein L650F, NH2-750F, Sulfate-650F)Kit per lo sviluppo del metodo8,00 mm100 mm5 ml1,3 ml/min (max. 2,5 ml/min)Log in per disponibilità1 * 1 STSFr. 1’840.00