Labels for thermal transfer printers
Thermal transfer printer labels for a variety of uses, including centrifuge, Eppendorf and PCR tube, conical and large tube, and vial laboratory applications.
- Polypropylene labels with permanent acrylic adhesive
- Gloss finish
- Thickness 0,076 mm
- Maximum service temperature is 120 °C
- Minimum service temperature is –196 °C
Thermal transfer labels are compatible with Brady printers BBP11, BBP12, PR200Plus, PR300Plus, PR600Plus, BP1244, BP1344, BP4000, and BP4320.
Thermal transfer printer labels for a variety of uses, including centrifuge, Eppendorf and PCR tube, conical and large tube, and vial laboratory applications.
- Polypropylene labels with permanent acrylic adhesive
- Gloss finish
- Thickness 0,076 mm
- Maximum service temperature is 120 °C
- Minimum service temperature is –196 °C
Thermal transfer labels are compatible with Brady printers BBP11, BBP12, PR200Plus, PR300Plus, PR600Plus, BP1244, BP1344, BP4000, and BP4320.
Product Family Options
- DescrizioneModelloMaterialeAdesivoPrinting methodColoreDimensioniConfezione
- Rectangle and circle printer labelsTHT-307-7600-1.5SCPPAcrilico permanenteThermal transfertrasparente9,53×25,40 mm1500 labels/roll1 * 1 ROUSFr. 198.00
- Rectangle printer labelsTHT-301-7600-2.5SCPPAcrilico permanenteThermal transfertrasparente9,53×25,40 mm2500 labels/roll1 * 1 ROUSFr. 300.00
- Rectangle printer labelsTHT-301-7600-3PPAcrilico permanenteThermal transfertrasparente9,53×25,40 mm3000 labels/roll1 * 1 ROUSFr. 297.00
- Rectangle and circle printer labelsTHT-304-7600-3PPAcrilico permanenteThermal transfertrasparente9,53×25,40 mm3000 labels/roll1 * 1 ROUSFr. 327.00
- Rectangle and circle printer labelsTHT-308-7600-1.5SCPPAcrilico permanenteThermal transfertrasparente12,70×25,40 mm1500 labels/roll1 * 1 ROUSFr. 216.00
- Rectangle and circle printer labelsTHT-309-7600-1.5SCPPAcrilico permanenteThermal transfertrasparente12,70×25,40 mm1500 labels/roll1 * 1 ROUSFr. 224.00
- Rectangle printer labelsTHT-310-7600-2.5SCPPAcrilico permanenteThermal transfertrasparente12,70×25,40 mm2500 labels/roll1 * 1 ROUSFr. 327.00
- Rectangle and circle printer labelsTHT-306-7600-3PPAcrilico permanenteThermal transfertrasparente12,70×25,40 mm3000 labels/roll1 * 1 ROUSFr. 356.00
- Rectangle printer labelsTHT-310-7600-10PPAcrilico permanenteThermal transfertrasparente12,70×25,40 mm10000 labels/pack1 * 10.000 STSFr. 998.00
- Rectangle printer labelsTHT-302-7600-1.5SCPPAcrilico permanenteThermal transfertrasparente15,24×41,28 mm1500 labels/roll1 * 1 ROUSFr. 257.00
- Rectangle printer labelsTHT-302-7600-3PPAcrilico permanenteThermal transfertrasparente15,24×41,28 mm3000 labels/roll1 * 1 ROUSFr. 427.00
- Square printer labelsTHT-305-7600-2.5SCPPAcrilico permanenteThermal transfertrasparente25,40×25,40 mm2500 labels/roll1 * 1 ROUSFr. 421.00
- Square printer labelsTHT-305-7600-3PPAcrilico permanenteThermal transfertrasparente25,40×25,40 mm3000 labels/roll1 * 1 ROUSFr. 427.00
- Rectangle printer labelsTHT-303-7600-1.5SCPPAcrilico permanenteThermal transfertrasparente25,40×48,26 mm1500 labels/roll1 * 1 ROUSFr. 196.00
- Rectangle printer labelsTHT-303-7600-3PPAcrilico permanenteThermal transfertrasparente25,40×48,26 mm3000 labels/roll1 * 1 ROUSFr. 327.00
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..