General purpose gloves, Showa 7166R
Catalog # 112-5892
Supplier: Showa
Specifications
- Dimensione del guanto:10
- Ambidestro/mano specifico:mano specifico
- Monouso/riutilizzabile:riutilizzabile
- Colore:blu scuro
- Lunghezza:260 mm
- Modello:7166R
- Norma EN:EN 388:2016 (4111X)
- Tipo polsino:Polsino di sicurezza
- Materiale rivestimento interno:Cotone jersey
- Finitura:Full nitrile coating
- Conf:1 PAIR
Specifications
About this item
The 7166 general purpose gloves feature an impermeable full nitrile coating that protects hands from a range of hazards, including abrasion, oils, and chemicals.
- Reinforced safety cuff prevents liquid and debris from entering the glove
- Soft cotton liner controls the temperature and wicks away sweat
- Rough surface grip is ideal for wet or greasy environments and increased protection
Typical applications include: Chemical spray and treatment, cutting roses and thorns, public utility, washing and cleaning, drainage, piping, guttering and drain pipe, plumbing, water/sewage treatment.
Certifications: EN 388:2016 (4111X); Cat. II