Cut resistant gloves, Showa GP-KV1
Nome del fornitore: Showa
Certificati
Informazioni su questo articolo
Showa GP-KV1 gloves are made of aramid fibre, flexible and easy-maintenance providing effective protection against cuts and with excellent dry and wet grip.
- Rough finish to latex coated palm
- Excellent mechanical performance
- Protects the hand in damp environments
- Breathable back of hand to reduce perspiration
- Designed for easy movement and continuous wear
- Seamless liner prevents irritation
- Antibacterial and anti-odour treatment
Ideal for use in aerospace, automotive, construction, manufacturing, municipal services, warehousing and distribution. Typical applications includes glass handling, masonry, bodywork, fabrication, recycling and tiling.
Certifications: EN 388:2016 (3X43C); Cat. II
Packaging: 10 pairs per polybag.
Specifiche
- Catalog No:
- 112-6001
- 112-6000
- 112-5999
- 112-6002
- Modello:
- GP-KV1
- GP-KV1
- GP-KV1
- GP-KV1
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Materiale rivestimento interno:
- Kevlar®
- Kevlar®
- Kevlar®
- Kevlar®
- Finitura:
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Latex coating on palm and fingertips
- Calibro:
- 10
- 10
- 10
- 10
- Tipo polsino:
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Maglia elastica
- Norma EN:
- EN 388:2016 (3X43C)
- EN 388:2016 (3X43C)
- EN 388:2016 (3X43C)
- EN 388:2016 (3X43C)