Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Ionic strength adjustor buffer solution for fluoride electrode, 1,4% HOAc, 8,2% NaOAc, 5,8% NaCl, trace CDTA), Plastic bottle, 475 mL
Codice catalogo 443123-475ML
Nome del fornitore: Merck
Ionic strength adjustor buffer solution for fluoride electrode, Supelco®
Codice catalogo 443123-475ML
Nome del fornitore: Merck
Numero CAS:
Vai a
Product Details & Documents
It may be used as TISAB buffer in determination of fluoride in coal, coal fly ash, phosphate rock, limestone, mineral clay, fossilised materials, oyster tissue and vegetation using pyrohydrolysis for sample decomposition.
Specifiche
Specifiche
- Tipo di conf.:Plastic bottle
- Conf:475 mL
- Descrizione:Ionic strength adjustor buffer solution for fluoride electrode
- Indice:1,4% HOAc, 8,2% NaOAc, 5,8% NaCl, trace CDTA)