ContactUsLinkComponent
Avantor® Hichrom, Unions
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
Avantor® Hichrom, Unions
Catalog # HICHHI-089
Supplier:  Avantor
undefined
Avantor® Hichrom, Unions
Catalog # HICHHI-089
Supplier:  Avantor
Specifications
  • Filettatura:
    6-32
  • Descrizione:
    True ZDV union
  • Materiale:
    PEEK
  • Seq:
    01
  • Conf:
    1
Specifications
About this item
These unions are made of PEEK.