Pharmaceutical secondary standard, Phenol, certified reference material, Supelco®
PHR1047-1G
: Merck
:
- Concentrazione:Neat
- Conf:1 g
- Standard:Pharmaceutical secondary standard (CRM)
- CAS No.:108-95-2
- Descrizione:Phenol
- Descrizione:Phenol
Traceable to USP 1524806.
Phenol may be used as a standard in determining the concentration of phenol present in samples of water using gas chromatography coupled with electron capture detector (GC-ECD).
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.