Supelco® Organic Reference Standard, Digoxin
Digoxin, Certified reference material, TraceCERT®, Bottomless glass bottle, 50 mg
Catalog # 04599-50MG
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS06
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P260 - P264 - P270 - P301 + P310 - P304 + P340 + P311 - P314
Specifications
Specifications
- Conf:50 mg
- Standard:Certified reference material, TraceCERT®
- Descrizione:Digoxin
- Pk Info:Bottomless glass bottle
- Peso molecolare:780,94
- Cat. No.:04599-50MG
- M-Clarity quality level:MQ300
- Condizioni di conservazione:15...25 °C
- Descrizione classe di pericolo:H300 - H331 - H373
- N. MDL:MFCD00003674
- Condizioni di spedizione:STD
- Synonym(s):12β-Hydroxydigitoxin
- CAS No.:20830-75-5



