ContactUsLinkComponent
VWR®, External PT100 Probe for Chillers
VWR®, External PT100 Probe for Chillers
Catalog # 462-0553
Supplier:  VWR Collection
VWR®, External PT100 Probe for Chillers
Catalog # 462-0553
Supplier:  VWR Collection
Specifications
  • Descrizione:
    External PT 100 probe for chillers, with 8 m cable length
  • Conf:
    1
Specifications
About this item
External PT100 probe is used for with chillers and coolers, 6000 series chiller, non-refrigerated coolers, 4100 liquid-to-liquid coolers.