Orders LinkContactUsLinkComponent
Laboratory coats
Laboratory coats
With elastic wrist, verde
Codice catalogo HOIT112849
Nome del fornitore:   HUBEI ORIENT INTL TRADING
Laboratory coats
Codice catalogo HOIT112849
Nome del fornitore:  HUBEI ORIENT INTL TRADING
Laboratory coats
Laboratory coats
With elastic wrist, verde
Price
SFr. 2.90
Log in per disponibilità
1 * 1 ST
Product Details & Documents
These disposable laboratory coats are ideal for non critical applications.
  • With elastic wrist
Weight: 40 g/m²
Colour: Blue/green.
Specifiche
  • Colore:
    verde
  • Descrizione:
    With elastic wrist
  • Seq:
    02
  • Cat. No.:
    HOIT112849
  • Peso:
    40 g/m²
  • Applicazioni per abiti:
    Laboratory