Laboratory coats, PP, with snap closure
These disposable laboratory coats made from PP material.
- With shirt, Korean, knitted collar
- Snaps for closures
Weight: 25 to 55 g/m²
Colour: White.
: 5 pieces/ bag, 10 bags/case.
- Catalog No:
- HOIT114527
- HOIT114525
- HOIT114526
- Genere:
- Unisex
- Unisex
- Unisex
- Applicazioni per abiti:
- Laboratory
- Laboratory
- Laboratory
- Peso:
- 25 - 55 g/m²
- 25 - 55 g/m²
- 25 - 55 g/m²
Frequently Bought Together