Avantor® Alltima HP C8, HPLC Columns, 5 µm
: Avantor
To overcome separation needs, these columns have high performance and selectivity.
- High purity silica
- Excellent column stability
- pH stability: 1 to 10
- Contains low retention then C18 phases
Used for reverse phase application where C18 is too rentention.
- Catalog No:
- HICH22465
- 87756/N
- HICH87751
- HICH87752
- Con endcapping:
- Sì
- Sì
- Sì
- Sì
- Codice USP:
- L7
- L7
- L7
- L7
Frequently Bought Together