Nucleosil C8, HPLC Columns
Catalog # NC100-7C8-10C5
Supplier: Avantor
Specifications
- Formato della colonna:Precartuccia
- Ø int.:3,20 mm
- Dimensioni particelle:7 µm
- Dimensioni pori:100 Å
- Lunghezza:10 mm
- Seq:38
- Fase:C8
- Conf:5
Specifications
About this item
Packed with classical spherical porous silica available in a wide range of pore sizes. The 100 Å and 120 Å have similar relative retention characteristics, however the 100 Å has the greater capacity whilst the 120 Å exhibits a lower backpressure.
- Good mechanical stability
- Good batch to batch reproducibility