Sigma Pseudo™ narcotic scent marijuana formulation, Supelco®
Catalog # P7309-100G
Supplier: Merck
CAS Number:
Specifications
- Conf:100 g
- Descrizione:Sigma Pseudo™ Narcotic Scent Marijuana formulation
- Descrizione:Sigma Pseudo(TM) Narcotic Scent Marijuana formulation
Specifications
About this item
Substitute for controlled substances in the training of narcotics detector dogs. Mimics the odor of marijuana.
Sigma pseudo™ narcotic scent marijuana formulation may be used as a surrogate in the comparative study of odor profiles of real marijuana odor vs surrogate marijuana scent using multidimensional gas chromatography coupled with mass spectrometry-olfactometry (MDGC– MS-O).
Caution: Storage temperature: −20 °C