Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standard
Standard organici
Enrofloxacin, analytical standard, VETRANAL™, Supelco®
Enrofloxacin, analytical standard, VETRANAL™, Supelco®
33699-100MG-R
:
Merck
:
Enrofloxacin, analytical standard, VETRANAL™, Supelco®
33699-100MG-R
:
Merck
:
Conf:
100 mg
Standard:
analytical standard
CAS No.:
93106-60-6
Descrizione:
Enrofloxacin
M-Clarity quality level:
MQ100
Descrizione:
Enrofloxacin