SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Standard
Standard organici
Lanoconazole, analytical standard, VETRANAL™, Supelco®
Lanoconazole, analytical standard, VETRANAL™, Supelco®
32991-25MG
:
Merck
:
Lanoconazole, analytical standard, VETRANAL™, Supelco®
32991-25MG
:
Merck
:
Conf:
25 mg
Standard:
analytical standard
CAS No.:
101530-10-3
Descrizione:
Lanoconazole
M-Clarity quality level:
MQ100
Descrizione:
Lanoconazole