Sulfate standard for IC, TraceCERT®, Supelco®
Référence 79735-100ML
Fournisseur: Merck
Numéro CAS:
Spécifications
- Concentrazione:10000 mg/l
- Conf:100 mL
- Descrizione:Sulphate in water
Spécifications
À propos de cet article
Certifications: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.