Orders LinkContactUsLinkComponent
Nitrate nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
N as nitrate in water, 1000 mg/l, 100 mL
Codice catalogo 53638-100ML
Nome del fornitore:   Merck
Nitrate nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
Codice catalogo 53638-100ML
Nome del fornitore:  Merck
Numero CAS:  
Nitrate nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
Nitrate nitrogen standard for IC, certified reference material, TraceCERT®, Supelco®
N as nitrate in water, 1000 mg/l, 100 mL
Numero CAS:  
Price
SFr. 81.20
Log in per disponibilità
1 * 100 mL
Product Details & Documents
 : This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.
Specifiche
Specifiche
  • Concentrazione:
    1000 mg/l
  • Conf:
    100 mL
  • Descrizione:
    N as nitrate in water
Specifiche