Multi-element Ion chromatography cation standard solution, certified, Supelco®
Multi-element Ion Chromatography Cation Standard Solution, 50 mL
Codice catalogo 89316-50ML-F
Nome del fornitore: Merck
Multi-element Ion chromatography cation standard solution, certified, Supelco®
Codice catalogo 89316-50ML-F
Nome del fornitore: Merck
Numero CAS:
Vai a
Product Details & Documents
10,0 mg/kg ±0,2% Li+, Na+, K+, Mg²⁺, Ca²⁺ each cation, certified reference material traceable to SI.
Specifiche
Specifiche
- Conf:50 mL
- Descrizione:Multi-element Ion Chromatography Cation Standard Solution
- Cat. No.:89316-50ML-F
- M-Clarity quality level:MQ300