ContactUsLinkComponent
Supelco® Organic Reference Standard, N-Propionylglycine
Supelco® Organic Reference Standard, N-Propionylglycine
Catalog # 92986-10MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, N-Propionylglycine
Catalog # 92986-10MG
Supplier:  Merck
CAS Number:  
Specifications
  • Conf:
    10 mg
  • Standard:
    Analytical standard
  • CAS No.:
    21709-90-0
  • Descrizione:
    N-Propionylglycine
  • Seq:
    01
  • Condizioni di conservazione:
    2…8 °C
  • Condizioni di spedizione:
    WICE
  • N. MDL:
    MFCD09049674
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2-(Propionylamino)acetic acid; N-(1-Oxopropyl)glycine; NSC 158539; Propionyl glycine
  • Peso molecolare:
    131,13