Pint bottle brush
114 mm, 63,5 mm, Plastica, 305 mm
Codice catalogo MICRBRWH-66
Nome del fornitore: MICRONOVA
Vai a
Product Details & Documents
This wirewound brush features a smaller diameter handle and softer bristles for delicate cleaning of jars and bottles with smaller openings.
- BPA free
Base resins used to produce block and bristles are made from FDA approved materials.
Specifiche
- Testa della spazzola Ø:63,5 mm
- Handle material:Plastica
- Lunghezza totale:305 mm
- Longitud del cabezal del cepillo:114 mm