Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 SFr.
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Vassoi/Piatti/Contenitori
Vassoi/piatti per laboratorio
Sample pans for Pyris 1 TGA and TGA 8000
Sample pans for Pyris 1 TGA and TGA 8000
Sample pans for Pyris 1 TGA and TGA 8000
Catalog # PERCN5370464
Supplier:
PerkinElmer
Expand image
Sample pans for Pyris 1 TGA and TGA 8000
Catalog # PERCN5370464
Supplier:
PerkinElmer
Specifications
Per:
TGA 8000 and Pyris 1 TGA
Descrizione:
Ceramic sample pans, pack of 10
Conf:
1
Specifications
About this item
Product Details
Product Details