EPA 552 halogenated acetic acids mix, certified reference material, Supelco®
Catalog # 48047.
Supplier: Merck
CAS Number:
Specifications
- Concentrazione:2000 mug/mL each component in methyl tert-butyl ether
- Conf:1
- Descrizione:EPA 552 Halogenated Acetic Acids Mix
- Environmentally Preferable:
- Tipo:Certified reference material