Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Pentadecafluorooctanoic acid ammonium salt
Supelco® Organic Reference Standard, Pentadecafluorooctanoic acid ammonium salt
Catalog # 91967-50MG
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Pentadecafluorooctanoic acid ammonium salt
Catalog # 91967-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Conf:
    50 mg
  • Standard:
    Analytical standard
  • CAS No.:
    3825-26-1
  • Descrizione:
    Pentadecafluorooctanoic acid ammonium salt
  • Environmentally Preferable:
  • Condizioni di conservazione:
    2...30 °C
  • Descrizione classe di pericolo:
    H302 - H318 - H331 - H351 - H360 - H362 - H372
  • Condizioni di spedizione:
    STD
  • N. MDL:
    MFCD00042599
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Ammonium pentadecafluorooctanoate; Perfluorocaprylic acid ammonium salt; Perfluorooctanoic acid ammonium salt
  • Formula:
    CF₃(CF₂)₆COONH₄
  • Peso molecolare:
    431,10

Specifications

About this item

  • GHS signal: GHS06, GHS08, GHS05
Caution: P260 - P263 - P280 - P304 + P340 + P311 - P305 + P351 + P338 - P308 + P313