Organic reference standard, Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate, analytical standard, Supelco®
93777-25MG
: Merck
:
- Conf:25 mg
- Standard:Analytical standard
- CAS No.:302776-68-7
- Descrizione:Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate
- Environmentally Preferable:
- Formula:C₂₄H₃₁NO₄
- Descrizione:Hexyl 2-[4-(diethylamino)-2-hydroxybenzoyl]benzoate
: Bottomless glass bottles are inserted with fused cone.