Organic reference standard, Ethylene glycol diacrylate, analytical standard, Supelco®
Catalog # 41608-100MG
Supplier: Merck
CAS Number:
Specifications
- Conf:100 mg
- Standard:Analytical standard
- CAS No.:2274-11-5
- Descrizione:Ethylene glycol diacrylate
- Formula:C₈H₁₀O₄
- Descrizione:Ethylene glycol diacrylate
- Punto di ebollizione:66...68 °C/2 mmHg(lit.)