Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, Bromodichloroacetic acid, analytical standard, Supelco®
Catalog # 442499.
Supplier:  Merck
CAS Number:  
Organic reference standard, Bromodichloroacetic acid, analytical standard, Supelco®
Catalog # 442499.
Supplier:  Merck
CAS Number:  

Specifications

  • Conf:
    1
  • Standard:
    Analytical standard
  • CAS No.:
    71133-14-7
  • Descrizione:
    Bromodichloroacetic acid
  • Punto di fusione:
    69...72 °C (lit.)
  • Formula:
    BrCl₂CCOOH
  • Temperatura di conservazione:
    room temprature
  • Descrizione:
    Bromodichloroacetic acid

Specifications

About this item