Mailing boxes, EPS
Nome del fornitore: DAKLAPACK
Vai a
Informazioni su questo articolo
Made up of expanded polystyrene (EPS).
- Non transparent
- Can be used for cooling elements, gel packs and perfect for dry ice
Suitable for refrigerated transport of biological substances, pharmaceuticals, and blood products.
4 varianti
Product Details & Documents
Made up of expanded polystyrene (EPS).
- Non transparent
- Can be used for cooling elements, gel packs and perfect for dry ice
Suitable for refrigerated transport of biological substances, pharmaceuticals, and blood products.
Product Family Options
Informazioni sul prodotto
- Esterno L×P×AInterno LxPxADisponibilitàPrice
- DAKL520005EPS insulation box with lid, 3 LEPS insulation box with lid, 3 L230×235×152 mm170×175×80 mmLog in per disponibilità1 * 1 ST24.60 SFr.
- DAKL13004EPS insulation box with lid, 12 LEPS insulation box with lid, 12 L250×340×320 mm180×270×250 mmLog in per disponibilità1 * 1 ST26.60 SFr.
- DAKL13005EPS insulation box with lid, 23 LEPS insulation box with lid, 23 L317×418×315 mm257×358×250 mmLog in per disponibilità1 * 1 ST28.20 SFr.
- DAKL520006Protective cardboard outer box, for 3 L boxProtective cardboard outer box, for 3 L box244×259×162 mm242×240×133 mmLog in per disponibilità1 * 1 ST24.70 SFr.