ContactUsLinkComponent
Microscope stand, rolling, Dino-Lite MS-W1
Microscope stand, rolling, Dino-Lite MS-W1
Codice catalogo IDCPMS-W1
Nome del fornitore:  Dino-Lite / IDCP B.V.
undefined
Microscope stand, rolling, Dino-Lite MS-W1
Codice catalogo IDCPMS-W1
Nome del fornitore:  Dino-Lite / IDCP B.V.
Specifiche
  • Descrizione:
    Rolling stand for Dino-Lite microscopes, MS-W1, ideal for surface inspections or for use on cylindrical surfaces
  • Seq:
    915
  • Conf:
    1
Specifiche
Informazioni su questo articolo
The MS-W1 stand with HD-M1 holster is a special stand for cylindrical surfaces or to roll on flat surfaces.
The arcs on the MS-W1 enable the user to easily examine cylindrical objects such as printing rolls. The barrel rollers at the bottom of the arcs are wrapped in rubber which helps to prevent the stand from scratching or harming the surfaces it is examining. The MS-W1 can be used to examine tubular objects of at least 7 centimeters in diameter. With the two barrel rollers, the MS-W1 stand can also be rolled forward or backward making it a great solution for surface inspection; such as inspection of artwork, blank PC boards, or any other flat surfaces. The HD-M1 holster included with the MS-W1 roller rack can be separated and be attached to poles 10 mm or 8 mm in diameter. The Dino-Lite is secured in place on the holster by a single thumbscrew. The holster is secured to a guidance pole also with a separate thumbscrew.
Compatible with all Dino-Lite models.