Five-fold tap banks, Discofix® C
Catalog # BRAU16609C
Supplier: B. Braun
Specifications
- Colore:blu
- Descrizione:Five-fold tap bank, drug-resistant
- Conf:40
Specifications
About this item
Five-fold tap banks are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppressants and lipids.
- Leaks caused by stress cracks, contamination and air embolism are prevented
- Easy access through the rotary adapter for injections and aspirations
- Control of the 45° position by 'clicking'
- Pressure resistant up to 2 bar
- PVC and DEHP-free