Three-fold multi-tap bank, Discofix® C
Katalognr. BRAU16615C
Anbieter: B. Braun
Technische Daten
- Colore:blu
- Descrizione:Three fold tap banks, drug-resistant
- Lunghezza:150 cm
- Conf:100
Technische Daten
Über diesen Artikel
Drug-Resistant Multi-Tap Systems are used for administration of medicines and medicines such as anesthetics, cytostatics, antibiotics, narcoleptics, immunosuppressants and lipids.
- Leaks caused by stress cracks, contamination and air embolism are prevented
- Easy access through the rotary adapter for injections and aspirations
- Control of the 45° position by 'clicking'