Tourniquets, easy
Catalog # PRAE904
Supplier: Praemeta
Specifications
- Descrizione:Tourniquet, 904, easy, red
- Conf:1
Specifications
About this item
Tourniquets made of latex-free, skin-friendly, durable cotton fabric.
- Suitable for fast blood sampling
- High quality metal lock in a modern design
- Separately and precisely adjustable decongestion
- Reduced tissue acidosis during extended use
Suitable for doctor's bags and first-aid kits.