Laboratory tray, PP
Nome del fornitore: MULTIROIR
Vai a
Informazioni su questo articolo
Range of five lightweight and sturdy laboratory trays. They are made of white heavy-duty PP and are ideal for a wide range of uses.
- Food grade
- Nestable
- Thermal resistance –25 to +70 °C
- Machine washable
Smooth easy to clean hygienic internal surface.
5 varianti
Product Details & Documents
Range of five lightweight and sturdy laboratory trays. They are made of white heavy-duty PP and are ideal for a wide range of uses.
- Food grade
- Nestable
- Thermal resistance –25 to +70 °C
- Machine washable
Smooth easy to clean hygienic internal surface.
Product Family Options
Informazioni sul prodotto
- CapacitàMaterialeL×P×AEsterno L×P×AInterno LxPxADisponibilitàPrice
- 2,8 L vol. (2 kg load)PP bianco305×195×60 mm245×145×55 mmLog in per disponibilità1 * 1 STSFr. 5.50Specifiche:
More
Specifications
Cat. No.216-2239 - Specifiche:
More
Specifications
Cat. No.216-2240 - Specifiche:
More
Specifications
Cat. No.216-2241 - Specifiche:
More
Specifications
Cat. No.216-2242 - 15,5 L (10 kg load)PP bianco545×400×86 mm ext.470×330×80 mmLog in per disponibilità1 * 12 STSFr. 201.00Specifiche:
More
Specifications
Cat. No.MULC1187-12