Microplates, opaque OptiPlates™
Marketsource
Nome del fornitore: PerkinElmer
Vai a
Informazioni su questo articolo
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
3 varianti
Product Details & Documents
These plates are used for general isotopic, fluorescence, LANCE™, [FP]², AlphaScreen™ or luminescence assays.
- Made of polystyrene
Product Family Options
Informazioni sul prodotto
- N. di pozzettiColoreConfezioneDisponibilitàPrice
- PERK6055270OptiiPlate™-96 F microplatesSpecifiche:MarketsourceMarketsource
- PERK6005290OptiiPlate™-96 microplatesSpecifiche:MarketsourceMarketsource
- PERK6005299OptiiPlate™-96 microplatesSpecifiche:MarketsourceMarketsource