Transfection reagent, in-vitro, TurboFect™
Catalog # R0532
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Conf:1 KIT
- DNA:Yes
- Descrizione:TurboFect™ transfection reagent, 1 ml
Specifications
About this item
TurboFect™ transfection reagent is recommended for efficient transfection of plasmid DNA. Distinct from lipid-based formulas, TurboFect™ is a sterile solution of a proprietary cationic polymer in water.
- Excellent transfection efficiency in the presence or absence of serum
- Ready-to-use reagent - no need to reconstitute, dilute, or manipulate
- Simple protocol
The polymer forms positively-charged complexes with DNA that are both compact and stable. These complexes protect DNA from degradation and facilitate efficient plasmid delivery into eukaryotic cells.
High transfection efficiency of plasmid DNA into a wide variety of cell types, that is adherent and suspension cells and differentiated and undifferentiated cells.
Caution: For research use only. Not for use in diagnostic procedures.