Dispensing pins, MICRO PIN®
Codice catalogo BRAU415019
Nome del fornitore: B. Braun
Specifiche
- Descrizione:MICRO PIN® non–vented, sterile dispensing pin
- Conf:100
Specifiche
Informazioni su questo articolo
Single use dispensing pin is used for withdrawal or injection of medication from rubber-stopper vials and bag injection ports.
- Compatible with Luer lock connection
- Simple design with large internal lumen
- Eliminates the risk of needle stick injuries
- High fluid-flow rate and comfortable easy handling
Made of plastic and free from PVC, DEHP and natural rubber latex.