VWR®, Filling and Venting Cap for VWR® Carboys
216-1842
- Descrizione:Filling/venting 3-port cap for VWR® carboys
- Conf:1
The filling and venting cap for VWR® carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water and chemicals
Each cap is equipped with integrated 6,35 mm ports.