Deuterium lamp for UV/Vis detector
Catalog # PERCN2920149
Supplier: PerkinElmer
Specifications
- Per:LC 200/785A
- Descrizione:Deuterium lamp
- Environmentally Preferable:
- Conf:1
Specifications
About this item
Deuterium lamp for the LC series 200 UV/Vis detector, provide outstanding performance and lamp life.
Lamp changes are quick and easy due to a unique, self-aligning lamp mount.