Chemical pads
Codice catalogo PRTWSM80YER
Nome del fornitore: Portwest
Specifiche
- Descrizione:Chemical pads, yellow
- L×L:500×400 mm
- Conf:200
Specifiche
Informazioni su questo articolo
Perforated single spun bonded layers gives excellent tensile strength and rapid absorption properties.
- Suitable for use in lint free environments
- Each pad can absorb 0,4 L of aggressive and non-aggressive chemicals
Certifications: Meets standards BS 7959-1 and BS 7959-3.